Learn More
Sino Biological Recombinant Human Siglec-2/CD22 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 11958H08H100
Encompass_Preferred
A DNA sequence encoding the human CD22 isoform-beta (P20273-1) extracellular domain (Met 1-Arg 687) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7645