Learn More
Sino Biological Recombinant Human ROBO4 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11960H08H100
Encompass_Preferred
A DNA sequence encoding the human ROBO4 (Q8WZ75-1) N-terminal four domains (Met 1-Glu 469) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7652