Learn More
eMolecules​ 1196156-51-0 | 4-BROMO-1,1-DIFLUOROCYCLOHEXANE | AstaTech | MFCD13189572 | 199.039 | C6H9BrF2 | 95.000 | FC1(F)CCC(Br)CC1 | 0.25g | 302810469
Catalog No. 5000493008 Shop All eMolecules Inc. Products
Encompass_Preferred
4-BROMO-1,1-DIFLUOROCYCLOHEXANE | AstaTech | 1196156-51-0 | MFCD13189572 | 199.039 | C6H9BrF2 | 95.000 | FC1(F)CCC(Br)CC1 | 0.25g | 302810469
Catalog No. 50-004-93008 Supplier eMolecules​ Supplier No. 302810469
4-BROMO-1,1-DIFLUOROCYCLOHEXANE | AstaTech | 1196156-51-0 | MFCD13189572 | 199.039 | C6H9BrF2 | 95.000 | FC1(F)CCC(Br)CC1 | 0.25g | 302810469