Learn More
Sino Biological Recombinant Human DAPK1 Protein (aa 1-363, His & GST Tag)
Supplier:  Sino Biological 11966H20B50
Encompass_Preferred
A DNA sequence encoding the N-terminal segment of human DAPK1 (P53355-1) (Met 1-Leu 363) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-7661