Learn More
Sino Biological Recombinant Human RPE Protein (His Tag)
Supplier:  Sino Biological 11970H08H100
Encompass_Preferred
A DNA sequence encoding the human RPE isoform 1 (NP_954699.1) (Met 1-Arg 228) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7664