Learn More
eMolecules 1197033-19-4 | 5'-ODMT cEt N-Bz A Phosphoramidite (Amidite) | Medchem Express (US)899.986 | C49H54N7O8P | 98.000 | COc1ccc(cc1)C(OC[C@]12O[C@H]([C@H](O[C@H]1C)[C@@H]2OP(OCCC#N)N(C(C)C)C(C)C)n1cnc2c(NC(=O)c3ccccc3)ncnc12)(c1ccccc1)c1ccc(OC)cc1 | 100mg | 911533450
Shop All eMolecules Inc. Products