Learn More
Sino Biological Recombinant Human Beta-2 microglobulin Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11976H08H100
Encompass_Preferred
A DNA sequence encoding the human B2M (NP_004039.1) (Met 1-Met 119) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7678