Learn More
Sino Biological Recombinant Human HIF-1 alpha/HIF1A Protein (His Tag)
Supplier:  Sino Biological 11977H07E100
Encompass_Preferred
A DNA sequence encoding the human HIF1A isoform 1 (Q16665-1) N-terminal segment (Arg 575-Asn 826) was expressed, with a polyhistide tag at the N-terminus.
Catalog No. 50-161-7680