Learn More
Sino Biological Recombinant Human EGFL7 Protein (His Tag)
Supplier:  Sino Biological 11979H07B100
Encompass_Preferred
A DNA sequence encoding the mature form of human EGFL7 (NP_057299.1) (Tyr 24-Ser 273) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-7685