Learn More
Sino Biological Recombinant Human FANCA Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11980H07B100
Encompass_Preferred
A DNA sequence encoding the human FANCA (AAH08979.1) (Met 1-Cys 297) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-7688