Learn More
Sino Biological Recombinant Human KEAP1 Protein (His & GST & AVI Tag)
Supplier:  Sino Biological 11981H29B100
Encompass_Preferred
A DNA sequence encoding the human KEAP1 (Q14145) (Gln 2-Cys 624) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus linked by the AVI tag.
Catalog No. 50-161-7692