Learn More
Sino Biological Recombinant Human KEAP1 Protein, HPLC-verified
Supplier:  Sino Biological 11981HNCB100
Encompass_Preferred
A DNA sequence encoding the human KEAP1 (Q14145) (Gln2-Cys624) was expressed and purified with two additional amino acids (Gly & Pro ) at the N-terminus.
Catalog No. 50-161-7694