Learn More
Sino Biological Recombinant Human Ubiquitin Activating Enzyme E1/UBA1 Protein (His & GST Tag)
Catalog No. 501617697
Encompass_Preferred
A DNA sequence encoding the human UBA1 (NP_003325.2) (Ser 2-Arg 1058) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-7697 Supplier Sino Biological Supplier No. 11990H20B50
A DNA sequence encoding the human UBA1 (NP_003325.2) (Ser 2-Arg 1058) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.