Learn More
Sino Biological Recombinant Human c-Kit Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 11996H02H50
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human KIT (P10721-2) (Met1-Thr516) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-7699