Learn More
Sino Biological Recombinant Human c-Kit Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 11996H08H100
Encompass_Preferred
A DNA sequence encoding the human KIT isoform 2 (P10721-2) extracellular domain (Met 1-Thr 516) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7702