Learn More
Sino Biological Recombinant Human RET Protein (His Tag)
Supplier:  Sino Biological 11997H08H150
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human RET (P07949-1) (Met 1-Arg 635) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7707