Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

11alpha-Hydroxyprogesterone Acetate 98.0+%, TCI America™
SDP

Catalog No. H04991G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H04991G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H04991G Supplier TCI America Supplier No. H04991G

Chemical Identifiers

CAS 2268-98-6
Molecular Formula C23H32O4
Molecular Weight (g/mol) 372.51
MDL Number MFCD00021152
InChI Key IWRPVTXREVYBHT-ZQEATNLPSA-N
Synonym 11alpha-Acetoxypregn-4-ene-3,20-dione, 11alpha-Acetoxyprogesterone, 11alpha-Hydroxypregn-4-ene-3,20-dione Acetate
PubChem CID 247927
IUPAC Name (1S,3aS,3bS,9aR,9bS,10R,11aS)-1-acetyl-9a,11a-dimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-10-yl acetate
SMILES CC(=O)O[C@@H]1C[C@@]2(C)[C@@H](CC[C@@H]2C(C)=O)[C@@H]2CCC3=CC(=O)CC[C@]3(C)[C@@H]12

Specifications

Melting Point 177°C
Color White-Yellow
Quantity 1 g
Formula Weight 372.51
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 11alpha-Hydroxyprogesterone Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.