Learn More
TARGETMOL CHEMICALS INC 11BETA-HSD1-IN-1 5MG
SDP
Catalog No. 502581757
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 11(beta)-HSD1-IN-1 is an inhibitor of 11(beta)-hydroxydehydrogenase 1 (11(beta)-HSD1 IC50 52 nM) and used for the treatment of pain. purity: 99%
Catalog No. 50-258-1757 Supplier TARGETMOL CHEMICALS INC Supplier No. T101605MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 11(beta)-HSD1-IN-1 is an inhibitor of 11(beta)-hydroxydehydrogenase 1 (11(beta)-HSD1 IC50 52 nM) and used for the treatment of pain. purity: 99%