Learn More
Medchemexpress LLC 11beta-Hydroxyprogesterone | 600-57-7 | 5 MG
SDP
Catalog No. 5000420794
Encompass_Preferred
11beta-Hydroxyprogesterone is a potent inhibitor of 11β-Hydroxysteroid dehydrogenase. It activates the human mineralocorticoid receptor in COS-7 cells (ED50 of 10 nM) and exhibits agonist mineralocorticoid activity by activating the transiently expressed human mineralocorticoid receptor (hMR) in COS-7 cells. Its agonist activity is linked to contacts between its 11β-hydroxyl group and Asn770 within the hMR-ligand-binding domain.
  • Potent inhibitor of 11β-Hydroxysteroid dehydrogenase.
  • Activates human mineralocorticoid receptor in COS-7 cells with an ED50 of 10 nM.
  • Displays agonist mineralocorticoid activity.
  • Exhibits hypertensinogenic activity in rats, dependent on an intact adrenal and mineralocorticoid receptor activation.
  • Classified as an endogenous metabolite and a steroid.
Catalog No. 50-004-20794 Supplier Medchemexpress LLC Supplier No. HYN23375MG
11beta-Hydroxyprogesterone is a potent inhibitor of 11β-Hydroxysteroid dehydrogenase. It activates the human mineralocorticoid receptor in COS-7 cells (ED50 of 10 nM) and exhibits agonist mineralocorticoid activity by activating the transiently expressed human mineralocorticoid receptor (hMR) in COS-7 cells. Its agonist activity is linked to contacts between its 11β-hydroxyl group and Asn770 within the hMR-ligand-binding domain.
  • Potent inhibitor of 11β-Hydroxysteroid dehydrogenase.
  • Activates human mineralocorticoid receptor in COS-7 cells with an ED50 of 10 nM.
  • Displays agonist mineralocorticoid activity.
  • Exhibits hypertensinogenic activity in rats, dependent on an intact adrenal and mineralocorticoid receptor activation.
  • Classified as an endogenous metabolite and a steroid.