Learn More
Strem, An Ascensus Company CAS# 791616-62-1. 100mg. (11bR)-2,6-Bis[3,5-bis(trifluoromethyl)phenyl]-4-hydroxy-4-oxide-dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin, 98%, (99% ee).
SDP
Catalog No. NC1519371
Encompass
CAS# 791616-62-1. 100mg. (11bR)-2,6-Bis[3,5-bis(trifluoromethyl)phenyl]-4-hydroxy-4-oxide-dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin, 98%, (99% ee). . Molecular Weight: 772.5. Molecular Formula: C36H17F12O4P. Color/form: white to light-yellow pwdr. Strem# 15-1366. http://www.strem.com/catalog/v/15-1366/
Catalog No. NC1519371 Supplier Strem, An Ascensus Company Supplier No. 151366100MG
CAS# 791616-62-1. 100mg. (11bR)-2,6-Bis[3,5-bis(trifluoromethyl)phenyl]-4-hydroxy-4-oxide-dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin, 98%, (99% ee). . Molecular Weight: 772.5. Molecular Formula: C36H17F12O4P. Color/form: white to light-yellow pwdr. Strem# 15-1366. http://www.strem.com/catalog/v/15-1366/