Learn More
Cayman Chemical 11dehydro Thromboxane B2 Quan
SDP
Catalog No. 502887021
Encompass_Preferred
The 11-dehydro TXB2 Quant-PAK has been designed for the convenient, precise quantification of 11-dehydro TXB2 by GC- or LC-MS. It includes a 50 UG vial of 11-dehydro TXB2-d4 and a precisely weighed vial of unlabeled 11-dehydro TXB2, with the precise weight (1 mg) indicated on the vial. This unlabeled 11-dehydro TXB2 can be used to quantify the 11-dehydro TXB2-d4 standard by constructing a standard curve of peak intensity ratios (deuterated versus unlabeled).
Catalog No. 50-288-7021 Supplier Cayman Chemical Supplier No. 100068311 EA
The 11-dehydro TXB2 Quant-PAK has been designed for the convenient, precise quantification of 11-dehydro TXB2 by GC- or LC-MS. It includes a 50 UG vial of 11-dehydro TXB2-d4 and a precisely weighed vial of unlabeled 11-dehydro TXB2, with the precise weight (1 mg) indicated on the vial. This unlabeled 11-dehydro TXB2 can be used to quantify the 11-dehydro TXB2-d4 standard by constructing a standard curve of peak intensity ratios (deuterated versus unlabeled).