Learn More
Cayman Chemical 11-dehydro Thromboxane B2 ELISA Kit - Monoclonal, 96 Well
SDP
Catalog No. 502895448
Encompass_Preferred
Cayman’s 11-dehydro TXB2 ELISA Kit is a competitive assay that can be used for the quantification of 11-dehydro TXB2 in plasma, serum, urine, and other sample matrices. The assay has a range of 15.6-2,000 pg/ml, with a midpoint (50% B/B0) of 80-160 pg/ml, and a sensitivity (80% B/B0) of approximately 34 pg/ml.
Catalog No. 50-289-5448 Supplier Cayman Chemical Supplier No. 51951096 WELL
Cayman’s 11-dehydro TXB2 ELISA Kit is a competitive assay that can be used for the quantification of 11-dehydro TXB2 in plasma, serum, urine, and other sample matrices. The assay has a range of 15.6-2,000 pg/ml, with a midpoint (50% B/B0) of 80-160 pg/ml, and a sensitivity (80% B/B0) of approximately 34 pg/ml.