Learn More
Cayman Chemical 11dehydro Thromn B2 El 100ng
SDP
Catalog No. 502943900
Encompass_Preferred
A cardenolide with diverse biological activities inhibits N-acetyltransferase (IC50 ~1.2 UM) reduces maximal Na /K -ATPase pump activity and cell attachment and increases cell death in MDCK-C7 cells at 1 UM decreases the levels of SRC-3 in MCF-7 cells at 500 nM increases calcium efflux and sarcoplasmic reticulum calcium levels in a voltage-clamp assay using isolated and perfused guinea pig hearts at 50 UM
Catalog No. 50-294-3900 Supplier Cayman Chemical Supplier No. 419514100 NG
A cardenolide with diverse biological activities inhibits N-acetyltransferase (IC50 ~1.2 UM) reduces maximal Na /K -ATPase pump activity and cell attachment and increases cell death in MDCK-C7 cells at 1 UM decreases the levels of SRC-3 in MCF-7 cells at 500 nM increases calcium efflux and sarcoplasmic reticulum calcium levels in a voltage-clamp assay using isolated and perfused guinea pig hearts at 50 UM