Learn More
Medchemexpress LLC 11H-indeno[1,2-b]quinoxalin-11-one O-(2-furanylcarbonyl)oxime | 312538-03-7 | 99.4% | 341.32 g·mol⁻1 | C20H11N3O3 | 100 MG
SDP
Catalog No. 5000240908
Encompass_Preferred
IQ-3 is a small-molecule inhibitor of the c-Jun N-terminal kinase (JNK) family with preference for JNK3. It binds JNK1, JNK2, and JNK3 with reported Kd values of 0.24 μM, 0.29 μM, and 0.066 μM, respectively. Supplied as an off-white to light yellow solid, IQ-3 is used in research to probe JNK signaling in vitro and in vivo; solubility, purity, and storage recommendations are provided on the product datasheet.
  • Preference for JNK3 over JNK1 and JNK2.
  • Low-micromolar to sub-micromolar binding affinities for JNK isoforms.
  • High purity (99.4%).
  • Available as a solid and in DMSO solution for convenient dosing.
  • Soluble in DMSO (25 mg/mL); sonication recommended.
  • Powder storage at -20°C with defined solvent stability.
Catalog No. 50-002-40908 Supplier Medchemexpress LLC Supplier No. HY107600100MG
IQ-3 is a small-molecule inhibitor of the c-Jun N-terminal kinase (JNK) family with preference for JNK3. It binds JNK1, JNK2, and JNK3 with reported Kd values of 0.24 μM, 0.29 μM, and 0.066 μM, respectively. Supplied as an off-white to light yellow solid, IQ-3 is used in research to probe JNK signaling in vitro and in vivo; solubility, purity, and storage recommendations are provided on the product datasheet.
  • Preference for JNK3 over JNK1 and JNK2.
  • Low-micromolar to sub-micromolar binding affinities for JNK isoforms.
  • High purity (99.4%).
  • Available as a solid and in DMSO solution for convenient dosing.
  • Soluble in DMSO (25 mg/mL); sonication recommended.
  • Powder storage at -20°C with defined solvent stability.