Learn More
S2 Media 11mL RPMI Broth with 2% MOPS and Glucose in 16x105mm PP Tube with Screw Cap, 50/box
Catalog No. 502995974
Encompass_Preferred
This item has a minimum order quantity of 10 per supplier requirements.
RPMI Broth with 2% MOPS and Glucose is an enrichment broth utilized for the selection and cultivation of fungi, including Candida species. This medium can be used in support of antifungal susceptibility testing, biofilm formation and pathogenesis studies.
• Product meets CLSI reference methods and performance criteria.
• RPMI Broth should be used by trained professionals within a laboratory setting.
• For in vitro diagnostic (IVD) use.
• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
Catalog No. 50-299-5974 Supplier S2 Media Supplier No. 5424
This item has a minimum order quantity of 10 per supplier requirements.
RPMI Broth with 2% MOPS and Glucose is an enrichment broth utilized for the selection and cultivation of fungi, including Candida species. This medium can be used in support of antifungal susceptibility testing, biofilm formation and pathogenesis studies.
• Product meets CLSI reference methods and performance criteria.
• RPMI Broth should be used by trained professionals within a laboratory setting.
• For in vitro diagnostic (IVD) use.
• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.