Learn More
Ace Glass, Inc. 11mm Ace-Thred nylon bushing, -012 FETFE O-ring, accepts 9-10.5mm OD tubing
SDP
Catalog No. 509557807
Encompass_Preferred
11mm Ace-Thred nylon bushing connector for joining an Ace-Thred to a reduced-end tube. Bushing accepts 9-10.5mm O.D. tubing. Includes -012 FETFE o-ring.
Catalog No. 50-955-7807 Supplier Ace Glass, Inc. Supplier No. 750602
11mm Ace-Thred nylon bushing connector for joining an Ace-Thred to a reduced-end tube. Bushing accepts 9-10.5mm O.D. tubing. Includes -012 FETFE o-ring.