Learn More
Analytical Sales & Services 11mm, 12 x 32, 2mL Amber Glass Wide Mouth Crimp/Snap Vials with Marking Spot
Catalog No. 5000807268
Encompass
Advantage™ 11mm, 12 x 32, 2mL Volume Amber Glass Wide Mouth Crimp/Snap Vials with Marking Spot - for Agilent, Leap and Perkin Elmer Autosamplers
Catalog No. 50-008-07268 Supplier Analytical Sales & Services Supplier No. 11212MCASE
Advantage™ 11mm, 12 x 32, 2mL Volume Amber Glass Wide Mouth Crimp/Snap Vials with Marking Spot - for Agilent, Leap and Perkin Elmer Autosamplers