Learn More
Analytical Sales & Services 11mm, 12 x 32, 2mL Wide Mouth Amber Glass Crimp Vials
Catalog No. 5000804949
Encompass
Advantage™ 11mm, 12 x 32 2mL Volume Wide Mouth Amber Glass Crimp Vials - for Use with Agilent, Leap and Perkin Elmer Autosamplers
Catalog No. 50-008-04949 Supplier Analytical Sales & Services Supplier No. 11222CASE
Advantage™ 11mm, 12 x 32 2mL Volume Wide Mouth Amber Glass Crimp Vials - for Use with Agilent, Leap and Perkin Elmer Autosamplers