Learn More
Analytical Sales & Services 11mm, 12 x 32, 2mL Wide Mouth Clear Glass Crimp Vials with Marking Spots
Catalog No. 5000806016
Encompass
Advantage™ 11mm, 12 x 32 2mL Volume Wide Mouth Clear Glass Crimp Vials with Marking Spots - for Use with Agilent, Leap and Perkin Elmer Autosamplers
Catalog No. 50-008-06016 Supplier Analytical Sales & Services Supplier No. 11221MCASE
Advantage™ 11mm, 12 x 32 2mL Volume Wide Mouth Clear Glass Crimp Vials with Marking Spots - for Use with Agilent, Leap and Perkin Elmer Autosamplers