Learn More
Ace Glass, Inc. 11MM 2MM BORE ADAPTER 11MM 2MM BORE ADAPTER
SDP
Catalog No. 50972231
Encompass_Preferred
11mm adapter with drip tip for chemical type chromatography. Adapter has 2mm bore with 1:5 taper PTFE stopcock at bottom for controlling flow.
Catalog No. 50-972-231 Supplier Ace Glass, Inc. Supplier No. 583507
11mm adapter with drip tip for chemical type chromatography. Adapter has 2mm bore with 1:5 taper PTFE stopcock at bottom for controlling flow.