Learn More
Perkin Elmer US LLC 11mm Electronic Universal Decapper with Global Mains Plug Pack
Catalog No. 502111529
Encompass_Preferred
Whatever your need may be, PerkinElmer offers a wide range of crimping tools for your convenience. Our universal voltage, precision control, power crimpers with adjustable settings are designed to deliver hundreds of crimps on a single battery charge. Battery: LiFePO4 6.4 VoltRated 1500 Charge Cycles Tested to EN62133 and UN DOT Very low self-discharge Direct plug connection to board PTCs integral to battery Crimper can operate while battery is charging. Motor: Stall torque 1400 g cm.Control Buttons: Top-mounted keypad equal visibility and convenience for left- and right handed use. Compliance: Safety: Machinery Directive and EN/IEC 61010 EMC: IEC 61326-1:2006Electronics: 20 MHz processor with A/D measurement of battery voltage and motor current. Advanced charging circuit. Additional on-board safety circuitry. PWM for soft-start of motor.
Catalog No. 50-211-1529 Supplier Perkin Elmer US LLC Supplier No. N9304502
Whatever your need may be, PerkinElmer offers a wide range of crimping tools for your convenience. Our universal voltage, precision control, power crimpers with adjustable settings are designed to deliver hundreds of crimps on a single battery charge. Battery: LiFePO4 6.4 VoltRated 1500 Charge Cycles Tested to EN62133 and UN DOT Very low self-discharge Direct plug connection to board PTCs integral to battery Crimper can operate while battery is charging. Motor: Stall torque 1400 g cm.Control Buttons: Top-mounted keypad equal visibility and convenience for left- and right handed use. Compliance: Safety: Machinery Directive and EN/IEC 61010 EMC: IEC 61326-1:2006Electronics: 20 MHz processor with A/D measurement of battery voltage and motor current. Advanced charging circuit. Additional on-board safety circuitry. PWM for soft-start of motor.