Learn More
Ace Glass, Inc. 11MM GRADED SEALS 11MM GRADED SEALS
SDP
Catalog No. 50974595
Encompass
11mm quartz graded seals for connecting Pyrex or Kimax borosilicate glass to Quartz glass. Quartz length is 7.6cm, and borosilicate length is 10.2cm.
Catalog No. 50-974-595 Supplier Ace Glass, Inc. Supplier No. 845518
11mm quartz graded seals for connecting Pyrex or Kimax borosilicate glass to Quartz glass. Quartz length is 7.6cm, and borosilicate length is 10.2cm.