Learn More
Description
Use certified clean, low particle, low background vials for you high sensitivity LC/MS and GC/MS applications. They are fully lot-tested for 15 critical parameters. Thermo Scientific™ Clear 11mm Snap Top MS Certified Vials are available in 2mL volumes. Septa are PTFE/Clear Silicone, Pre-slit. Pre-cleaned vial packaging protects the product integrity and high purity closures packed in air-tight re-closeable container.
- Up to 15 critical physical characteristics affecting vial performance
- Low background by positive ESI LC/MS
- Low background by GC/MS
Specifications
Specifications
| Description | Convenience Kit, 11mm Snap Top Vial, 2mL |
| Color | Clear |
| Material | Glass |
| No. per Pack | 100 |
| Marking Spot | Yes |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.