Learn More
Chemglass Life Sciences Tube, Connecting, "T" Shaped, CG-300-02 (10mm Small Serration, 11mm Large Serration) Hose Connection
SDP
Catalog No. CG402202
Encompass_Preferred
Tube, Connecting, "T" Shaped, CG-300-02 (10mm Small Serration, 11mm Large Serration) Hose Connection. T shaped tube having serrated ends for connection of rubber, latex or Tygon tubing.
Catalog No. CG402202 Supplier Chemglass Life Sciences Supplier No. CG402202
Tube, Connecting, "T" Shaped, CG-300-02 (10mm Small Serration, 11mm Large Serration) Hose Connection. T shaped tube having serrated ends for connection of rubber, latex or Tygon tubing.