Learn More
SEPSOLUTIONS 11MM ORANGE SNAP CAPS 100/PK
Catalog No. NC9626595
Encompass_Preferred
11mm one piece orange snap cap with pre-inserted PTFE/silicone/PTFE solid septum with no slit, compatible with all 12x32mm snap/crimp vials incuding StepVials, pack of 100.
Catalog No. NC9626595 Supplier SEPSOLUTIONS Supplier No. 501318
11mm one piece orange snap cap with pre-inserted PTFE/silicone/PTFE solid septum with no slit, compatible with all 12x32mm snap/crimp vials incuding StepVials, pack of 100.