Learn More
Analytical Sales & Services 11mm Red Snap Caps with Silicone/PTFE Liners
Catalog No. 5000804274
Encompass
Advantage™ 11mm Red Snap Caps with Silicone/PTFE Liners
Catalog No. 50-008-04274 Supplier Analytical Sales & Services Supplier No. 20044RCASE
Advantage™ 11mm Red Snap Caps with Silicone/PTFE Liners