Learn More
GENTECH SOLUTIONS LLC 11mm Silver Aluminum Crimp Seal, .020" PTFE Disk Septum
Catalog No. NC3728813
Encompass_Preferred
EliteChem Made in the USA with the highest purity silicone and premium-quality PTFE films to produce the Septum in the closures. Compatible with chromatography vials with a 11mm seal diameter.
Catalog No. NC3728813 Supplier GENTECH SOLUTIONS LLC Supplier No. 2000210011
EliteChem Made in the USA with the highest purity silicone and premium-quality PTFE films to produce the Septum in the closures. Compatible with chromatography vials with a 11mm seal diameter.