Learn More
Analytical Sales & Services 11mm Snap Caps with PTFE/Natural Rubber Liners
Catalog No. 5000806436
Encompass
Advantage™ 11mm Snap Caps with PTFE/Natural Rubber Liners
Catalog No. 50-008-06436 Supplier Analytical Sales & Services Supplier No. 20043CASE
Advantage™ 11mm Snap Caps with PTFE/Natural Rubber Liners