Learn More
Ace Glass, Inc. 11mm adapter, PTFE, polyethylene packing slip bottom drip
SDP
Catalog No. 50972243
Encompass_Preferred
11mm bottom-drip, Luer-Lok, PTFE adapter with flow regulator valve for use with all #11 Ace- Thred chromatography columns. Bore is adjustable from 0-1mm max. Supplied with one FETFE O-Ring and one replaceable polyethylene packing support.
Catalog No. 50-972-243 Supplier Ace Glass, Inc. Supplier No. 583746
11mm bottom-drip, Luer-Lok, PTFE adapter with flow regulator valve for use with all #11 Ace- Thred chromatography columns. Bore is adjustable from 0-1mm max. Supplied with one FETFE O-Ring and one replaceable polyethylene packing support.