Learn More
Cayman Chemical 11-methyl Lauric Acid methyl ester, Purity: ≥98%, CAS: 5129-57-7, 10 mg
SDP
Catalog No. 503015980
Encompass_Preferred
A methylated fatty acid methyl ester; has been used as a standard in the quantification of 11-methyl lauric acid in vernix caseosa and M. oleifera extract
Catalog No. 50-301-5980 Supplier Cayman Chemical Supplier No. 2480910 MG
A methylated fatty acid methyl ester; has been used as a standard in the quantification of 11-methyl lauric acid in vernix caseosa and M. oleifera extract