Learn More
Cayman Chemical 11mthyl LaurIc ACD mthyl 20mg
SDP
Catalog No. 503015963
Encompass_Preferred
A methylated fatty acid methyl ester; has been used as a standard in the quantification of 11-methyl lauric acid in vernix caseosa and M. oleifera extract
Catalog No. 50-301-5963 Supplier Cayman Chemical Supplier No. 2480920 MG
Add to Cart
Add to Cart
A methylated fatty acid methyl ester; has been used as a standard in the quantification of 11-methyl lauric acid in vernix caseosa and M. oleifera extract