Learn More
USA Scientific Inc 11-place chest freezer rk
Catalog No. 502745115
Encompass
11-place (1 long x 11 high) chest freezer rack for 2 in H boxes, 5 1/2 in L x 5 5/8 in W x 24 3/16 in H, stainless steel
Catalog No. 50-274-5115 Supplier USA Scientific Inc Supplier No. 26021100
11-place (1 long x 11 high) chest freezer rack for 2 in H boxes, 5 1/2 in L x 5 5/8 in W x 24 3/16 in H, stainless steel