Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(11R,12R)-9,10-Dihydro-9,10-ethanoanthracene-11,12-diamine 98.0+%, TCI America™
SDP

Catalog No. D34441G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34441G 1 g
D3444100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D34441G Supplier TCI America Supplier No. D34441G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 181139-49-1
Molecular Formula C16H16N2
Molecular Weight (g/mol) 236.318
InChI Key NWDYSRZJOLDMRE-QDIHITRGSA-N
PubChem CID 2794493
SMILES C1=CC=C2C3C(C(C(C2=C1)C4=CC=CC=C34)N)N

Specifications

Melting Point 159°C
Color White-Yellow
Quantity 1 g
Formula Weight 236.32
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (11R,12R)-9,10-Dihydro-9,10-ethanoanthracene-11,12-diamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.