Learn More
Medchemexpress LLC 11R-VIVIT | 592517-80-1 | 99.6% | 3573.15 | 1 MG
SDP
Catalog No. 5000424578
Encompass_Preferred
11R-VIVIT is a cell-permeable nuclear factor of activated T cells (NFAT) inhibitor. It can be used for the research of podocyte and diabetic nephropathy.
  • Cell-permeable to inhibit NFAT activity
  • Specifically targets and inhibits nuclear factor of activated T cells
  • Useful for the study of podocyte and diabetic nephropathy
  • Reduces NFAT2 expression in high glucose-treated podocytes in vitro
  • Attenuates diabetic nephropathy symptoms and restores podocyte number in vivo
Catalog No. 50-004-24578 Supplier Medchemexpress LLC Supplier No. HYP14301MG
11R-VIVIT is a cell-permeable nuclear factor of activated T cells (NFAT) inhibitor. It can be used for the research of podocyte and diabetic nephropathy.
  • Cell-permeable to inhibit NFAT activity
  • Specifically targets and inhibits nuclear factor of activated T cells
  • Useful for the study of podocyte and diabetic nephropathy
  • Reduces NFAT2 expression in high glucose-treated podocytes in vitro
  • Attenuates diabetic nephropathy symptoms and restores podocyte number in vivo