Learn More
Cayman Chemical 11S 12R-EET 100UG
SDP
Catalog No. 502895940
Encompass_Preferred
An oxylipin and a metabolite of arachidonic acid; activates KCa1.1/BK in isolated rat coronary small arterial smooth muscle cells at 50 nM; induces dilation of precontracted isolated canine epicardial arterioles (EC50 = 20 pM) and denuded porcine subepicardial arterioles (EC50 = 10 pM)
Catalog No. 50-289-5940 Supplier Cayman Chemical Supplier No. 9004348100 UG
An oxylipin and a metabolite of arachidonic acid; activates KCa1.1/BK in isolated rat coronary small arterial smooth muscle cells at 50 nM; induces dilation of precontracted isolated canine epicardial arterioles (EC50 = 20 pM) and denuded porcine subepicardial arterioles (EC50 = 10 pM)