Learn More
Chemglass Life Sciences Graded Seal, Quartz to Borosilicate, 11mm ID X 13mm OD
SDP
Catalog No. CGQ031010
Encompass_Preferred
Graded Seal, Quartz to Borosilicate, 11mm ID X 13mm OD. Graded seals have 3 inches of quartz sealed to 4 inches of borosilicate.
Catalog No. CGQ031010 Supplier Chemglass Life Sciences Supplier No. CGQ031010
Graded Seal, Quartz to Borosilicate, 11mm ID X 13mm OD. Graded seals have 3 inches of quartz sealed to 4 inches of borosilicate.