Learn More
Ace Glass, Inc. 11X300ML ADDITION FUNNEL 11X300Ml Addition Funnel
SDP
Catalog No. 50972204
Encompass_Preferred
Addition funnels with Ace-Thred™ connections for a leak-proof seal. Match with corresponding column Ace-Thred™ and join with our 5840 or 5841 product family of couplings to create a connection with no significant reduction in I.D. Available in cylindrical or conical styles.
Catalog No. 50-972-204 Supplier Ace Glass, Inc. Supplier No. 582205
Addition funnels with Ace-Thred™ connections for a leak-proof seal. Match with corresponding column Ace-Thred™ and join with our 5840 or 5841 product family of couplings to create a connection with no significant reduction in I.D. Available in cylindrical or conical styles.