Learn More
Ace Glass, Inc. 11mm ID 300mm chromatography column with #11 Ace-Threds.
SDP
Catalog No. 50972180
Encompass_Preferred
11mm I.D. X 300mm rugged chromatography column with straight-through, internal #11 AceThred ends, for use as a packing column or as an operating column. Designed for high performance, low pressure liquid chromatography (HPLPLC).
Catalog No. 50-972-180 Supplier Ace Glass, Inc. Supplier No. 582004
11mm I.D. X 300mm rugged chromatography column with straight-through, internal #11 AceThred ends, for use as a packing column or as an operating column. Designed for high performance, low pressure liquid chromatography (HPLPLC).