Learn More
Cayman Chemical 12:0 N-Biotinyl Fatty Acid, Purity: ≥98%, CAS: 135447-73-3, 10 mg
SDP
Catalog No. 502932825
Encompass_Preferred
A biotinylated form of lauric acid; an intermediate in the synthesis of N-biotinylated phenylarsonous acids
Catalog No. 50-293-2825 Supplier Cayman Chemical Supplier No. 2571410 MG
A biotinylated form of lauric acid; an intermediate in the synthesis of N-biotinylated phenylarsonous acids