Learn More
AdipoGen 12-1-Pyrenyl dodecanoic acid
SDP
Supplier:  AdipoGen CDXP0006M005
Encompass_Preferred
Chemical. CAS 69168-45-2. Formula C28H32O2. MW 400.55. Synthetic. Anionic membrane probe used for studies on the uptake of fluorescent fatty acids into cultured cells and oxygen sensing.
Catalog No. 50-256-0290